Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178175.3 | NP_835469.1 | 4 | 946 | ||
XM_011530943.2 | XP_011529245.1 | 4 | 1156 | |||
XM_011530944.2 | XP_011529246.1 | 4 | 775 | |||
XM_011530946.2 | XP_011529248.1 | 3 | 878 | |||
XM_017029485.1 | XP_016884974.1 | 4 | 1351 | |||
GenBank mRNA | AK289946.1 | - | 4 | 979 | ||
AY217350.1 | - | 4 | 853 | |||
AY358208.1 | - | 3 | 755 | |||
BC100785.2 | - | 4 | 707 | 40000445 | ||
BC100786.1 | - | 3 | 746 | 40000448 | ||
BC100787.2 | - | 2 | 350 |