Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 4 | 772 | ||
XM_011519626.2 | XP_011517928.1 | 4 | 923 | |||
XM_011519627.2 | XP_011517929.1 | 4 | 649 | |||
XM_011519628.1 | XP_011517930.1 | 3 | 490 | |||
XM_011519629.1 | XP_011517931.1 | 3 | 538 | |||
XM_017016574.1 | XP_016872063.1 | 4 | 1498 | |||
GenBank mRNA | AK300456.1 | - | 3 | 515 | ||
BC157856.1 | - | 4 | 772 | 9057075 |