Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIA1 SIRNA Assays ›
Gene Symbol:
PPFIA1
Gene Name:
PTPRF interacting protein alpha 1
Gene Aliases:
LIP.1, LIP1, LIPRIN
Chromosome Location:
Chr.11: 70270687 - 70384501 on Build GRCh38
UniGene ID:
Hs.530749
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s507
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003626.3
NP_003617.1
22
3247
NM_177423.2
NP_803172.1
22
3247
NR_045286.1
23
3258
XM_006718716.3
XP_006718779.1
27
3572
XM_011545306.2
XP_011543608.1
27
3444
XM_011545307.2
XP_011543609.1
26
3545
XM_011545308.2
XP_011543610.1
26
3542
XM_011545309.2
XP_011543611.1
27
3572
XM_011545310.2
XP_011543612.1
26
3497
XM_011545311.2
XP_011543613.1
25
3485
XM_011545312.2
XP_011543614.1
25
3482
XM_011545313.2
XP_011543615.1
26
3455
XM_011545314.2
XP_011543616.1
25
3425
XM_011545315.2
XP_011543617.1
25
3380
XM_011545316.2
XP_011543618.1
24
3365
XM_011545317.2
XP_011543619.1
23
3335
XM_011545318.2
XP_011543620.1
12
1677
XM_011545319.1
XP_011543621.1
24
3353
XM_017018448.1
XP_016873937.1
24
3452
XM_017018449.1
XP_016873938.1
23
3290
GenBank mRNA
AB209314.1
-
23
3185
AK123331.1
-
1824
AK124621.1
-
2
1792
BC034046.1
-
22
3210
4794300
BX648037.1
-
21
2547
D49354.1
-
7
1019
U22815.1
-
22
3261
U22816.1
-
22
3261
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
signal transduction
neurotransmitter secretion
glutamate secretion
negative regulation of stress fiber assembly
negative regulation of establishment of protein localization to plasma membrane
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0