Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198498.2 | NP_940900.1 | 3 | 213 | ||
XM_011542803.2 | XP_011541105.1 | 4 | 356 | |||
XM_011542804.2 | XP_011541106.1 | 3 | 139 | |||
GenBank mRNA | BC039669.1 | - | 3 | 213 | 5167284 |