Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206920.1 | NP_001193849.1 | 4 | 784 | ||
NM_001306087.1 | NP_001293016.1 | 4 | 787 | |||
XM_011536720.2 | XP_011535022.1 | 6 | 4566 | |||
XM_011536721.2 | XP_011535023.1 | 6 | 4563 | |||
XM_011536723.2 | XP_011535025.1 | 4 | 1052 | |||
XM_011536724.2 | XP_011535026.1 | 4 | 1657 | |||
XM_011536725.1 | XP_011535027.1 | 5 | 976 | |||
XM_017021258.1 | XP_016876747.1 | 5 | 972 | |||
XM_017021259.1 | XP_016876748.1 | 6 | 4566 | |||
XM_017021260.1 | XP_016876749.1 | 5 | 979 | |||
XM_017021261.1 | XP_016876750.1 | 4 | 855 | |||
GenBank mRNA | BC101318.1 | - | 4 | 725 | ||
BC101319.1 | - | 4 | 1798 | |||
BC101320.1 | - | 4 | 731 | |||
BC101321.1 | - | 4 | 731 |