Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182971.2 | NP_892016.1 | 2 | 472 | ||
GenBank mRNA | AY161004.1 | - | 2 | 472 |