Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FKTN SIRNA Assays ›
Gene Symbol:
FKTN
Gene Name:
fukutin
Gene Aliases:
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
Chromosome Location:
Chr.9: 105558117 - 105655950 on Build GRCh38
UniGene ID:
Hs.55777
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5087
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FKTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079802.1
NP_001073270.1
5
510
NM_001198963.1
NP_001185892.1
5
510
NM_006731.2
NP_006722.2
4
418
XM_006717014.2
XP_006717077.1
4
422
XM_011518368.2
XP_011516670.1
6
658
XM_011518369.2
XP_011516671.1
5
508
XM_011518370.2
XP_011516672.1
6
2194
XM_011518371.2
XP_011516673.1
6
693
XM_011518373.2
XP_011516675.1
6
604
XM_011518374.2
XP_011516676.1
4
422
XM_011518375.2
XP_011516677.1
4
737
XM_011518376.2
XP_011516678.1
4
560
XM_011518378.2
XP_011516680.1
6
658
XM_011518379.2
XP_011516681.1
4
304
XM_011518381.2
XP_011516683.1
5
493
XM_011518387.2
XP_011516689.1
6
658
XM_011518389.2
XP_011516691.1
6
658
XM_011518391.2
XP_011516693.1
6
658
XM_017014462.1
XP_016869951.1
7
792
XM_017014463.1
XP_016869952.1
4
881
XM_017014464.1
XP_016869953.1
6
658
XM_017014465.1
XP_016869954.1
5
508
XM_017014466.1
XP_016869955.1
6
658
XM_017014467.1
XP_016869956.1
5
917
XM_017014468.1
XP_016869957.1
4
800
XM_017014469.1
XP_016869958.1
6
658
XM_017014470.1
XP_016869959.1
6
658
XM_017014471.1
XP_016869960.1
7
756
XM_017014472.1
XP_016869961.1
7
782
XM_017014473.1
XP_016869962.1
2
2660
XM_017014474.1
XP_016869963.1
4
421
XM_017014475.1
XP_016869964.1
4
425
XM_017014476.1
XP_016869965.1
6
658
XM_017014477.1
XP_016869966.1
5
523
XM_017014478.1
XP_016869967.1
7
725
XM_017014479.1
XP_016869968.1
4
422
GenBank mRNA
AB008226.1
-
4
405
AB609007.1
-
405
AK300840.1
-
5
508
AK304460.1
-
4
416
AL601876.1
-
4
421
BC101808.1
-
3
320
8069314
BC112038.1
-
3
320
8327506
BC117699.1
-
3
326
40073633
BC117700.1
-
4
450
40073634
DA917113.1
-
5
508
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nervous system development
muscle organ development
negative regulation of cell proliferation
protein O-linked mannosylation
negative regulation of JNK cascade
regulation of protein glycosylation
transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0