Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080532.1 | NP_001074001.1 | 19 | 2113 | ||
GenBank mRNA | AY236490.1 | - | 19 | 2191 | ||
AY263163.1 | - | 19 | 2152 | |||
BC132679.1 | - | 19 | 2150 | 40146701 | ||
BC143410.1 | - | 19 | 2150 | 9051917 | ||
BC157874.1 | - | 19 | 2162 | 9057092 |