Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013407.2 | NP_001013425.2 | 2 | 363 | ||
GenBank mRNA | BC101344.1 | - | 2 | 349 | 40025152 |
Gene Symbol: PRAMEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146344.2 | NP_001139816.2 | 2 | 340 | ||
XM_011541479.2 | XP_011539781.1 | 1 | 526 | |||
GenBank mRNA | BC144426.1 | - | 2 | 339 | 9052947 |
Gene Symbol: PRAMEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010889.2 | NP_001010889.1 | 2 | 349 | ||
GenBank mRNA | BC101342.2 | - | 2 | 349 | 40025149 |
Gene Symbol: PRAMEF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010890.2 | NP_001010890.2 | 1 | 269 |