Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1211L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207362.2 | NP_997245.2 | 7 | 2442 | ||
XM_005263923.3 | XP_005263980.1 | 7 | 2422 | |||
XM_005263924.3 | XP_005263981.1 | 7 | 2422 | |||
XM_005263926.3 | XP_005263983.1 | 7 | 2307 | |||
XM_011511093.1 | XP_011509395.1 | 9 | 2581 | |||
XM_011511094.1 | XP_011509396.1 | 8 | 2479 | |||
XM_011511095.2 | XP_011509397.1 | 7 | 2249 | |||
XM_011511096.1 | XP_011509398.1 | 4 | 1859 | |||
XM_017003971.1 | XP_016859460.1 | 7 | 2425 | |||
XM_017003972.1 | XP_016859461.1 | 8 | 2529 | |||
XM_017003973.1 | XP_016859462.1 | 7 | 2435 |