Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009565.1 | NP_001009565.1 | 8 | 815 | ||
XM_011532815.2 | XP_011531117.1 | 9 | 2231 | |||
XM_011532816.2 | XP_011531118.1 | 9 | 2231 | |||
XM_011532817.2 | XP_011531119.1 | 9 | 2108 | |||
XM_017003981.1 | XP_016859470.1 | 9 | 1583 | |||
XM_017003982.1 | XP_016859471.1 | 9 | 1059 | |||
GenBank mRNA | AY845084.1 | - | 8 | 815 | ||
AY847283.1 | - | 8 | 815 | |||
BC144460.1 | - | 9 | 988 |