Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198184.1 | NP_937827.1 | 1 | 80 | ||
XM_005247428.4 | XP_005247485.1 | 2 | 328 | |||
XM_017006303.1 | XP_016861792.1 | 2 | 239 | |||
GenBank mRNA | AY398681.1 | - | 1 | 80 | ||
AY573933.1 | - | 1 | 80 | |||
BC128106.1 | - | 1 | 80 | 40112103 | ||
BC128107.1 | - | 1 | 80 | 40112104 |