Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017992.3 | NP_001017992.1 | 1 | 658 | ||
GenBank mRNA | BC137470.1 | - | 1 | 576 | 9021470 | |
BC137473.1 | - | 1 | 576 | 9021473 | ||
BX648504.1 | - | 1 | 659 |