Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 5 | 370 | ||
NM_001167741.1 | NP_001161213.1 | 4 | 300 | |||
XM_005248495.3 | XP_005248552.1 | 5 | 372 | |||
XM_017009440.1 | XP_016864929.1 | 4 | 294 | |||
GenBank mRNA | AK296864.1 | - | 5 | 365 | ||
BC150295.1 | - | 5 | 360 | |||
BX538064.1 | - | 4 | 280 | |||
BX647596.1 | - | 5 | 367 | |||
DB456588.1 | - | 5 | 370 |