Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198511.2 | NP_940913.1 | 4, 5 | 1399 | ||
GenBank mRNA | AK124915.1 | - | 4, 5 | 1399 | ||
BC093667.1 | - | 4, 5 | 1191 | 7939512 | ||
BC093669.1 | - | 4, 5 | 1191 | 7939514 |