Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286509.1 | NP_001273438.1 | 4 | 1224 | ||
NM_001286510.1 | NP_001273439.1 | 4 | 1223 | |||
NM_001286511.1 | NP_001273440.1 | 4 | 1178 | |||
NM_001286512.1 | NP_001273441.1 | 4 | 1227 | |||
NM_001286513.1 | NP_001273442.1 | 3 | 1073 | |||
NM_001286517.1 | NP_001273446.1 | 3 | 1034 | |||
NM_001286519.1 | NP_001273448.1 | 2 | 879 | |||
NM_001286520.1 | NP_001273449.1 | 4 | 1232 | |||
NM_178148.3 | NP_835361.1 | 4 | 1228 | |||
GenBank mRNA | AB097021.1 | - | 5 | 1065 | ||
AB097039.1 | - | 5 | 1065 | |||
AB106538.1 | - | 4 | 1076 | |||
AK075456.1 | - | 4 | 1157 | |||
AK123749.1 | - | 4 | 1125 | |||
AK223028.1 | - | 3 | 878 | |||
AK293251.1 | - | 2 | 846 | |||
AK293344.1 | - | 3 | 1030 | |||
AK307572.1 | - | 4 | 1163 | |||
AY491520.1 | - | 4 | 1194 | |||
BC016839.1 | - | 1 | 251 | |||
BC024288.1 | - | 4 | 1192 | 4890949 |