Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKDD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182703.5 | NP_874362.3 | 12 | 1152 | ||
GenBank mRNA | BC047607.2 | - | 2 | 156 | ||
BC101274.1 | - | 9 | 850 | 40024168 | ||
BC101275.1 | - | 9 | 850 | 40024170 | ||
BC101276.1 | - | 9 | 843 |