Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 10 | 1216 | ||
NM_001271197.1 | NP_001258126.1 | 10 | 1296 | |||
NM_182619.3 | NP_872425.2 | 10 | 1322 | |||
XM_005255934.1 | XP_005255991.1 | 9 | 1265 | |||
XM_017023206.1 | XP_016878695.1 | 9 | 1265 | |||
XM_017023207.1 | XP_016878696.1 | 8 | 1095 | |||
XM_017023208.1 | XP_016878697.1 | 9 | 1448 | |||
XM_017023209.1 | XP_016878698.1 | 8 | 1158 | |||
GenBank mRNA | AF428259.1 | - | 9 | 1334 | ||
AF521893.1 | - | 10 | 1335 | |||
AK074773.1 | - | 9 | 1150 | |||
AK289884.1 | - | 9 | 1264 | |||
BC078143.1 | - | 10 | 1198 | 5761001 | ||
BC141808.1 | - | 10 | 1296 | 40147524 |
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 9 | 1278 | ||
XM_005255976.2 | XP_005256033.1 | 9 | 1355 | |||
XM_011523093.2 | XP_011521395.1 | 8 | 1061 | |||
XM_017023255.1 | XP_016878744.1 | 10 | 1532 | |||
XM_017023256.1 | XP_016878745.1 | 8 | 1248 | |||
GenBank mRNA | BM695392.1 | - | 5 | 489 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 9 | 1184 | ||
XM_006721179.1 | XP_006721242.1 | 9 | 1280 | |||
XM_011523034.1 | XP_011521336.1 | 10 | 1526 | |||
XM_011523036.2 | XP_011521338.1 | 8 | 1110 | |||
XM_011523037.1 | XP_011521339.1 | 8 | 1173 | |||
GenBank mRNA | AF448856.1 | - | 9 | 1334 | ||
AL833339.1 | - | 7 | 2282 | |||
BC039068.1 | - | 8 | 1966 | 5188643 |