Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010905.2 | NP_001010905.1 | 5 | 822 | ||
GenBank mRNA | AK303850.1 | - | 5 | 822 | ||
BC062712.1 | - | 5 | 826 | 30353154 |