Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181337.3 | NP_851854.1 | 1 | 1281 | ||
GenBank mRNA | AF181722.1 | - | 1 | 1281 |