Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181539.4 | NP_853517.2 | 5, 6 | 1017 | ||
GenBank mRNA | AJ564205.2 | - | 5, 6 | 1017 | ||
BC132951.1 | - | 5, 6 | 1008 | 40146973 | ||
BC136896.1 | - | 5, 6 | 1008 | 9020887 |