Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198514.3 | NP_940916.2 | 5 | 1266 | ||
XM_011539769.2 | XP_011538071.1 | 5 | 1266 | |||
GenBank mRNA | AK090631.1 | - | 5 | 1237 | ||
AK098487.1 | - | 3 | 380 | |||
BC032598.1 | - | 1 | 30 | 5529619 | ||
BX538287.1 | - | 1 | 74 |