Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C12ORF42 SIRNA Assays ›
Gene Symbol:
C12orf42
Gene Name:
chromosome 12 open reading frame 42
Gene Aliases:
-
Chromosome Location:
Chr.12: 103047669 - 103525258 on Build GRCh38
UniGene ID:
Hs.534649
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51559
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C12orf42
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001099336.2
NP_001092806.1
4
490
NM_001278419.1
NP_001265348.1
4
490
NM_001278420.1
NP_001265349.1
4
355
NM_198521.3
NP_940923.2
4
355
NR_103526.1
4
355
XM_006719377.3
XP_006719440.1
3
293
XM_011538291.1
XP_011536593.1
4
350
XM_011538292.2
XP_011536594.1
4
319
XM_011538293.2
XP_011536595.1
4
326
XM_011538294.2
XP_011536596.1
4
300
XM_011538295.1
XP_011536597.1
4
382
XM_011538297.2
XP_011536599.1
5
1366
XM_011538300.1
XP_011536602.1
5
535
XM_011538305.1
XP_011536607.1
4
552
XM_011538306.1
XP_011536608.1
4
350
XM_011538308.2
XP_011536610.1
5
380
XM_011538312.2
XP_011536614.1
5
378
XM_011538315.2
XP_011536617.1
5
371
XM_017019276.1
XP_016874765.1
4
444
XM_017019277.1
XP_016874766.1
7
819
XM_017019278.1
XP_016874767.1
6
598
XM_017019279.1
XP_016874768.1
7
1124
XM_017019280.1
XP_016874769.1
7
824
XM_017019281.1
XP_016874770.1
5
375
GenBank mRNA
AK058052.1
-
4
313
BC039352.1
-
4
313
BC044617.1
-
4
451
5267756
BC117392.1
-
4
313
40125943
BC117394.1
-
4
313
40125945
BI830750.1
-
4
432
HY002043.1
-
4
448
HY015666.1
-
4
355
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0