Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198525.2 | NP_940927.2 | 16, 17 | 3387 | ||
XM_011521531.2 | XP_011519833.1 | 16, 17 | 3497 | |||
GenBank mRNA | AY358384.1 | - | 12, 13 | 1956 | ||
BC040878.1 | - | 4, 5 | 686 | |||
BC104044.1 | - | 12, 13 | 1929 | 40026959 | ||
BC112271.1 | - | 11, 12 | 1820 | 8327739 | ||
BC112273.1 | - | 11, 12 | 1820 | 8327741 | ||
BC143461.1 | - | 11, 12 | 1816 | 9051969 |