Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP9B SIRNA Assays ›
Gene Symbol:
ATP9B
Gene Name:
ATPase phospholipid transporting 9B (putative)
Gene Aliases:
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location:
Chr.18: 79069275 - 79378391 on Build GRCh38
UniGene ID:
Hs.465475
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51592
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP9B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306085.1
NP_001293014.1
20
2432
NM_198531.4
NP_940933.3
20
2432
XM_011525963.2
XP_011524265.1
21
2676
XM_011525964.2
XP_011524266.1
20
2391
XM_011525965.2
XP_011524267.1
21
2672
XM_011525966.2
XP_011524268.1
20
2627
XM_011525971.2
XP_011524273.2
14
4734
XM_011525972.2
XP_011524274.1
15
1751
XM_011525973.2
XP_011524275.1
13
1525
XM_011525974.2
XP_011524276.1
13
1547
XM_017025726.1
XP_016881215.1
21
2676
XM_017025727.1
XP_016881216.1
21
2676
XM_017025728.1
XP_016881217.1
21
2598
XM_017025729.1
XP_016881218.1
21
2676
XM_017025730.1
XP_016881219.1
21
2598
XM_017025731.1
XP_016881220.1
21
2676
XM_017025732.1
XP_016881221.1
21
2598
XM_017025733.1
XP_016881222.1
21
2598
XM_017025734.1
XP_016881223.1
21
2676
XM_017025735.1
XP_016881224.1
21
2676
XM_017025736.1
XP_016881225.1
21
2676
XM_017025737.1
XP_016881226.1
21
2676
XM_017025738.1
XP_016881227.1
17
1908
XM_017025739.1
XP_016881228.1
17
1926
XM_017025740.1
XP_016881229.1
16
1831
XM_017025741.1
XP_016881230.1
18
2171
GenBank mRNA
AK127634.1
-
4
293
AK308291.1
-
20
2314
BC125219.1
-
20
2310
40123958
BC125220.1
-
20
2310
40123959
BX537860.1
-
18
1815
BX648948.1
-
8
1007
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
endocytosis
phospholipid translocation
establishment of protein localization to Golgi
magnesium ion binding
phospholipid-translocating ATPase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0