Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198534.2 | NP_940936.2 | 3 | 749 | ||
XM_005272473.3 | XP_005272530.1 | 3 | 920 | |||
XM_011527993.1 | XP_011526295.1 | 3 | 749 | |||
XM_011527995.2 | XP_011526297.1 | 3 | 749 | |||
GenBank mRNA | AK093103.1 | - | 3 | 734 | ||
BC029824.1 | - | 3 | 709 | 5170600 | ||
BC034779.1 | - | 3 | 698 | 5168945 |