Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198566.3 | NP_940968.1 | 9 | 1843 | ||
XM_011514038.2 | XP_011512340.1 | 9 | 1752 | |||
XM_017009445.1 | XP_016864934.1 | 8 | 1874 | |||
XM_017009446.1 | XP_016864935.1 | 7 | 1784 | |||
GenBank mRNA | AK056925.1 | - | 9 | 1758 | ||
BC036867.1 | - | 9 | 1706 | 5198039 |