Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198572.2 | NP_940974.2 | 4 | 1490 | ||
XM_011517021.1 | XP_011515323.1 | 4 | 1490 | |||
XM_011517022.2 | XP_011515324.1 | 4 | 1776 | |||
XM_011517023.2 | XP_011515325.1 | 4 | 2794 | |||
XM_011517024.2 | XP_011515326.1 | 5 | 2015 | |||
XM_011517025.2 | XP_011515327.1 | 4 | 2450 | |||
XM_017013399.1 | XP_016868888.1 | 5 | 1309 | |||
GenBank mRNA | AB092352.1 | - | 4 | 1478 | ||
BC050390.1 | - | 4 | 1480 | 5742710 | ||
BC053547.1 | - | 4 | 1465 | 5743874 |