Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGFR2 SIRNA Assays ›
Gene Symbol:
FGFR2
Gene Name:
fibroblast growth factor receptor 2
Gene Aliases:
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location:
Chr.10: 121478330 - 121598656 on Build GRCh38
UniGene ID:
Hs.533683
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5175
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGFR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000141.4
NP_000132.3
18
2984
NM_001144914.1
NP_001138386.1
15
2151
NM_001144916.1
NP_001138388.1
15
2433
NM_001144917.1
NP_001138389.1
16
2636
NM_001144918.1
NP_001138390.1
16
2633
NM_001320654.1
NP_001307583.1
17
2368
NM_001320658.1
NP_001307587.1
18
2978
NM_022970.3
NP_075259.4
18
2987
NM_023029.2
NP_075418.1
16
2220
NR_073009.1
17
2787
XM_006717708.2
XP_006717771.1
18
3179
XM_006717710.3
XP_006717773.1
18
3277
XM_006717711.2
XP_006717774.1
16
2223
XM_006717712.2
XP_006717775.1
15
2145
XM_017015920.1
XP_016871409.1
18
3271
XM_017015921.1
XP_016871410.1
18
3268
XM_017015922.1
XP_016871411.1
16
2217
XM_017015923.1
XP_016871412.1
16
2214
XM_017015924.1
XP_016871413.1
16
2837
XM_017015925.1
XP_016871414.1
16
2923
GenBank mRNA
AB084153.1
-
15
2001
AK308621.1
-
15
2433
BC037338.2
-
17
2368
BC039243.1
-
16
2633
5277896
EF057068.1
-
16
2070
M55614.1
-
17
2352
M80634.1
-
18
2758
M87770.1
-
18
2610
M87771.1
-
16
2476
M97193.1
-
18
2951
S41878.1
-
2
164
X52832.1
-
18
2516
Z71929.1
-
17
2349
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
angiogenesis
ureteric bud development
in utero embryonic development
epithelial to mesenchymal transition
positive regulation of mesenchymal cell proliferation
outflow tract septum morphogenesis
membranous septum morphogenesis
apoptotic process
cell-cell signaling
axonogenesis
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
post-embryonic development
embryonic pattern specification
organ morphogenesis
regulation of cell fate commitment
positive regulation of phospholipase activity
regulation of phosphatidylinositol 3-kinase signaling
morphogenesis of embryonic epithelium
peptidyl-tyrosine phosphorylation
orbitofrontal cortex development
ventricular zone neuroblast division
pyramidal neuron development
gland morphogenesis
positive regulation of Wnt signaling pathway
bone mineralization
lung development
epithelial cell differentiation
midbrain development
otic vesicle formation
hair follicle morphogenesis
lacrimal gland development
regulation of osteoblast proliferation
multicellular organism growth
organ growth
fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
fibroblast growth factor receptor signaling pathway involved in hemopoiesis
fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of fibroblast growth factor receptor signaling pathway
inner ear morphogenesis
odontogenesis
positive regulation of MAPK cascade
positive regulation of GTPase activity
cell fate commitment
regulation of osteoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lung alveolus development
mesodermal cell differentiation
embryonic digestive tract morphogenesis
embryonic organ morphogenesis
digestive tract development
embryonic organ development
reproductive structure development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
epidermis morphogenesis
branching morphogenesis of a nerve
mesenchymal cell differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
regulation of smooth muscle cell differentiation
positive regulation of cell division
ventricular cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
limb bud formation
bone development
bone morphogenesis
branching involved in prostate gland morphogenesis
branching involved in salivary gland morphogenesis
bud elongation involved in lung branching
lung lobe morphogenesis
lung-associated mesenchyme development
positive regulation of epithelial cell proliferation involved in lung morphogenesis
prostate gland morphogenesis
prostate epithelial cord elongation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
lateral sprouting from an epithelium
mammary gland bud formation
epithelial cell proliferation involved in salivary gland morphogenesis
branch elongation involved in salivary gland morphogenesis
branching involved in labyrinthine layer morphogenesis
regulation of branching involved in prostate gland morphogenesis
regulation of morphogenesis of a branching structure
mesenchymal cell differentiation involved in lung development
mesenchymal cell proliferation involved in lung development
regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0