Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199183.2 | NP_954652.2 | 4 | 585 | ||
GenBank mRNA | BC133695.1 | - | 4 | 585 | 40126663 |