Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199352.3 | NP_955384.3 | 1 | 235 | ||
XM_017017684.1 | XP_016873173.1 | 4 | 1314 | |||
XM_017017685.1 | XP_016873174.1 | 5 | 1442 | |||
XM_017017686.1 | XP_016873175.1 | 4 | 1750 | |||
XM_017017687.1 | XP_016873176.1 | 4 | 1855 | |||
XM_017017695.1 | XP_016873184.1 | 4 | 1855 | |||
GenBank mRNA | AK127610.1 | - | 1 | 110 | ||
AY437532.1 | - | 1 | 235 | |||
BC101315.1 | - | 1 | 230 | |||
BC101316.1 | - | 1 | 235 | |||
BC101317.1 | - | 1 | 235 |