Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031853.4 | NP_001027024.3 | 6 | 801 | ||
NM_001042536.2 | NP_001036001.1 | 6 | 725 | |||
NM_001278313.1 | NP_001265242.1 | 5 | 687 | |||
NM_001278314.1 | NP_001265243.1 | 6 | 792 | |||
NM_001278315.1 | NP_001265244.1 | 5 | 687 | |||
NM_001278316.1 | NP_001265245.1 | 5 | 687 | |||
XM_006718227.3 | XP_006718290.1 | 7 | 854 | |||
XM_011520085.1 | XP_011518387.1 | 6 | 729 | |||
XM_011520086.1 | XP_011518388.1 | 6 | 729 | |||
XM_017017697.1 | XP_016873186.1 | 7 | 766 | |||
XM_017017698.1 | XP_016873187.1 | 6 | 729 | |||
GenBank mRNA | AB231745.1 | - | 5 | 681 | ||
AB231747.1 | - | 5 | 681 | |||
AB231748.1 | - | 2 | 182 | |||
AB236158.1 | - | 6 | 725 | |||
AB236159.1 | - | 6 | 801 | |||
BC127700.1 | - | 5 | 687 | 40127962 | ||
BU147420.1 | - | 6 | 712 |