Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306084.1 | NP_001293013.1 | 65 | 9119 | ||
XM_011539069.2 | XP_011537371.1 | 66 | 9319 | |||
XM_011539070.2 | XP_011537372.1 | 66 | 9319 | |||
XM_011539071.2 | XP_011537373.1 | 66 | 9277 | |||
XM_011539072.2 | XP_011537374.1 | 66 | 9319 | |||
XM_011539073.2 | XP_011537375.1 | 66 | 9319 | |||
XM_011539074.2 | XP_011537376.1 | 66 | 9160 | |||
XM_011539076.2 | XP_011537378.1 | 66 | 9118 | |||
XM_011539077.2 | XP_011537379.1 | 65 | 9469 | |||
XM_011539079.1 | XP_011537381.1 | 62 | 8565 | |||
XM_011539085.1 | XP_011537387.1 | 51 | 7140 | |||
XM_017018866.1 | XP_016874355.1 | 66 | 9319 | |||
XM_017018867.1 | XP_016874356.1 | 55 | 7571 | |||
GenBank mRNA | CF887548.1 | - | 1 | 116 |