Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012754.3 | NP_001012772.1 | 2 | 430 | ||
NM_001017370.2 | NP_001017370.1 | 2 | 430 | |||
NR_073109.1 | 2 | 172 | ||||
GenBank mRNA | AK299198.1 | - | 2 | 343 | ||
AK300260.1 | - | 2 | 343 | |||
AL833329.1 | - | 1 | 847 | |||
BC052492.1 | - | 2 | 110 | |||
BC140877.1 | - | 2 | 356 | 9021745 | ||
BC140878.1 | - | 2 | 356 | 9021746 | ||
BX647872.1 | - | 2 | 320 | |||
BX648122.1 | - | 2 | 382 | |||
CR749318.1 | - | 2 | 351 | |||
DA772703.1 | - | 2 | 172 |