Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010978.3 | NP_001010978.2 | 5 | 484 | ||
NM_001276392.1 | NP_001263321.1 | 3 | 303 | |||
NM_001276393.1 | NP_001263322.1 | 4 | 355 | |||
NM_001276394.1 | NP_001263323.1 | 3 | 217 | |||
NM_001276395.1 | NP_001263324.1 | 4 | 324 | |||
XM_011541446.1 | XP_011539748.1 | 5 | 464 | |||
XM_017001264.1 | XP_016856753.1 | 4 | 307 | |||
GenBank mRNA | BC103740.1 | - | 3 | 205 | ||
BC127708.1 | - | 5 | 481 | 40127476 | ||
BC127709.1 | - | 3 | 300 | 40127477 | ||
BC140760.1 | - | 4 | 299 | 9021628 | ||
BC144466.1 | - | 4 | 299 | 9052992 | ||
BC171743.1 | - | 4 | 268 |