Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207398.2 | NP_997281.2 | 4, 5 | 576 | ||
GenBank mRNA | AK096141.1 | - | 4, 5 | 576 |