Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143962.1 | NP_001137434.1 | 6, 7 | 899 | ||
XM_017001265.1 | XP_016856754.1 | 3, 4 | 340 | |||
XM_017001267.1 | XP_016856756.1 | 6, 7 | 899 | |||
XM_017001268.1 | XP_016856757.1 | 6, 7 | 899 | |||
GenBank mRNA | BC157893.1 | - | 6, 7 | 836 | 9057098 |