Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207644.2 | NP_997527.2 | 4 | 1053 | ||
GenBank mRNA | AK097054.1 | - | 4 | 2769 | ||
BC017595.1 | - | 3 | 1301 | |||
BC032388.1 | - | 4 | 1014 | |||
BC108684.1 | - | 4 | 1055 | 6471196 | ||
DB535827.2 | - | 1 | 543 |