Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320493.1 | NP_001307422.1 | 2 | 519 | ||
NM_016206.3 | NP_057290.2 | 2 | 519 | |||
XM_006713138.3 | XP_006713201.1 | 1, 2 | 686 | |||
GenBank mRNA | BC094780.1 | - | 2 | 515 | 30528902 | |
CF995043.1 | - | 542 | ||||
CR936700.1 | - | 2 | 156 |