Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320493.1 | NP_001307422.1 | 2 | 673 | ||
NM_001320494.1 | NP_001307423.1 | 1, 2 | 514 | |||
NM_016206.3 | NP_057290.2 | 2 | 673 | |||
XM_006713138.3 | XP_006713201.1 | 2 | 840 | |||
GenBank mRNA | BC094780.1 | - | 2 | 669 | 30528902 | |
CR936700.1 | - | 2 | 310 |