Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFEM1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021485.2 | 2 | 625 | |||
GenBank mRNA | BI461452.1 | - | 2 | 487 | ||
DR000794.1 | - | 2 | 632 |