Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPL10 SIRNA Assays ›
Gene Symbol:
RPL10
EEF1A1
Gene Name:
ribosomal protein L10
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
AUTSX5, DXS648, DXS648E, L10, NOV, QM
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.X: 154398065 - 154402339 on Build GRCh38
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.534404
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s52451
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPL10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256577.2
NP_001243506.2
4
514
NM_001303624.1
NP_001290553.1
3
309
NM_001303625.1
NP_001290554.1
4
518
NM_001303626.1
NP_001290555.1
3
184
NM_006013.4
NP_006004.3
4
514
GenBank mRNA
AB019572.1
-
2
106
AF218023.1
-
2
1073
AF486812.1
-
1
183
AK026568.1
-
3
306
AK027197.1
-
1
1089
AK130582.1
-
4
202
AK307511.1
-
4
201
BC003358.1
-
3
185
2900175
BC026276.1
-
1
202
4276627
BC071918.1
-
1
180
6731213
BC095425.1
-
1
192
BF667333.1
-
4
349
BM761549.1
-
4
206
BX647954.1
-
1
202
CA776692.1
-
2
146
CR456797.1
-
3
161
CR542069.1
-
3
161
DQ369703.1
-
3
161
DQ369704.1
-
3
161
DQ369705.1
-
3
161
DQ369706.1
-
3
161
DQ369707.1
-
3
161
DQ369708.1
-
3
161
DQ369709.1
-
3
161
DQ369710.1
-
3
161
DQ369711.1
-
3
161
DQ369712.1
-
3
161
DQ369713.1
-
3
161
DQ369714.1
-
3
161
DQ369715.1
-
3
161
DQ369716.1
-
3
161
F30590.1
-
1
37
KJ534934.1
-
3
161
KJ534979.1
-
3
161
M64241.1
-
4
202
M73791.1
-
3
175
S35959.1
-
2
139
S35960.1
-
1
180
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK292639.1
-
202
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031100, XM_574109, X87106, FQ229696, FQ220555, and others.
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
structural constituent of ribosome
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0