Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024613.3 | NP_001019784.2 | 3 | 1065 | ||
NM_001160264.2 | NP_001153736.1 | 4 | 915 | |||
XM_005250337.3 | XP_005250394.1 | 4 | 2736 | |||
XM_011516202.2 | XP_011514504.1 | 5 | 2586 | |||
GenBank mRNA | AK300742.1 | - | 3 | 1194 | ||
AY726588.1 | - | 3 | 986 | |||
BC127714.1 | - | 3 | 1053 | 40127896 | ||
BC127715.1 | - | 4 | 903 | 40127897 | ||
BC136684.1 | - | 3 | 1121 | 9020674 | ||
BC144367.1 | - | 3 | 1121 | 9052888 |