Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289973.1 | NP_001276902.1 | 6 | 1079 | ||
NM_207411.4 | NP_997294.3 | 5 | 941 | |||
XM_017013401.1 | XP_016868890.1 | 4 | 1360 | |||
XM_017013402.1 | XP_016868891.1 | 3 | 1152 | |||
GenBank mRNA | AY358489.1 | - | 5 | 812 | ||
AY534242.1 | - | 5 | 941 | |||
AY534243.1 | - | 6 | 933 |