Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303531.1 | NP_001290460.1 | 1 | 68 | ||
NM_207412.2 | NP_997295.1 | 1 | 68 | |||
GenBank mRNA | AK125570.1 | - | 1 | 68 | ||
BC137511.1 | - | 1 | 63 | 9021511 | ||
BC137512.1 | - | 1 | 63 | 9021512 | ||
BC139744.1 | - | 1 | 63 | 40107968 |