Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015038.2 | NP_001015038.1 | 3 | 237 | ||
XM_011530785.2 | XP_011529087.1 | 4 | 653 | |||
XM_011530786.2 | XP_011529088.1 | 3 | 320 | |||
XM_011530787.2 | XP_011529089.1 | 3 | 300 | |||
XM_017029513.1 | XP_016885002.1 | 3 | 257 | |||
GenBank mRNA | BC130359.1 | - | 3 | 196 | 40146390 | |
BM782405.1 | - | 3 | 222 |