Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000204.4 | NP_000195.2 | 2 | 509 | ||
NM_001318057.1 | NP_001304986.1 | 2 | 509 | |||
XM_005262976.1 | XP_005263033.1 | 2 | 509 | |||
XM_006714209.1 | XP_006714272.1 | 2 | 509 | |||
XM_006714210.3 | XP_006714273.1 | 2 | 509 | |||
XM_011531920.1 | XP_011530222.1 | 2 | 509 | |||
XM_017008164.1 | XP_016863653.1 | 2 | 509 | |||
XM_017008165.1 | XP_016863654.1 | 2 | 509 | |||
XM_017008166.1 | XP_016863655.1 | 2 | 509 | |||
GenBank mRNA | AK290625.1 | - | 2 | 282 | ||
AK299232.1 | - | 2 | 335 | |||
BC020718.1 | - | 2 | 324 | 4716122 | ||
BP235907.1 | - | 2 | 462 | |||
DC298508.1 | - | 2 | 509 | |||
DC392360.1 | - | 2 | 463 | |||
J02770.1 | - | 2 | 269 | |||
KU159429.1 | - | 2 | 489 | |||
Y00318.1 | - | 2 | 284 |