Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR8H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005200.1 | NP_001005200.1 | 1 | 198 |
Gene Symbol: OR8H3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005201.1 | NP_001005201.1 | 1 | 198 |