Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258338.1 | NP_001245267.1 | 3 | 422 | ||
NM_018838.4 | NP_061326.1 | 4 | 510 | |||
GenBank mRNA | AF112208.1 | - | 4 | 530 | ||
BC005936.1 | - | 4 | 499 | 4069306 | ||
BG943528.1 | - | 4 | 495 | |||
BM548072.1 | - | 577 | ||||
BU532909.1 | - | 3 | 409 | |||
U34343.1 | - | 616 |