Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLNC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127487.1 | NP_001120959.1 | 30 | 5305 | ||
NM_001458.4 | NP_001449.3 | 30 | 5305 | |||
GenBank mRNA | AB208865.1 | - | 7 | 947 | ||
AB371585.1 | - | 30 | 5325 | |||
AF089841.1 | - | 30 | 5145 | |||
AF146692.1 | - | 30 | 5308 | |||
AJ012737.1 | - | 30 | 5114 | |||
X70084.1 | - | 2 | 187 |